Learn More
Ampac™ Flexibles SealPAK Pouches
Ideal for histology and pathology specimens and for safe biological waste disposal in medical, institutional and industrial facilities
Supplier:  Ampac™ 400 24
 View more versions of this product
Catalog No. 0181225A
Only null left
Description
Description
Ideal for histology and pathology specimens and for safe biological waste disposal in medical, institutional and industrial facilities
Recommended for applications requiring long-term storage, sharper objects, or chemical solutions.
  • Transparent pouches with polyethylene lining
  • Can withstand infrared and gamma rays
  • May be frozen, microwaved, or boiled without harm
  • Easily resealable
  • 2.5 mil
  • Use with Ampac Flexibles pouch sealer (Cat. Nos. 01-812-15 or 01-812-15B) and handy pouch cutter (01-812-21)
Order Info
Also available in heat-sealable tubular roll stock that allows you to create custom-size pouches.
Specifications
Chemical Identifiers
-56°C to +115°C
2.50 mil
Specifications
PET, Polyethylene
-56°C to +115°C
6 x 4 in. (15.2 x 10.2cm)
4 in.
15.2 cm
2400/Cs.
2.50 mil
6 in.
4 oz.
10.2 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories