missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ AMP PNP
Description
AMP PNP is a non-hydrolyzable AMP analog. Kir6 (KATP) channel blocker. Inhibits fast axonal transport and stabilizes the interaction of membranous organelles with microtubules.
Specifications
Specifications
| CAS | 72957-42-7 |
| Quantity | 10 mg |
| Solubility | Soluble to 50mM in water |
| Product Line | Tocris |
| Molecular Formula | C10H13Li4N6O12P3 |
| Formula Weight | 529.93g/mol |
| Color | White |
| Product Type | Inhibitor |
For Research Use Only