missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amodiaquin Dihydrochloride Dihydrate 98.0+%, TCI America™
Supplier: TCI America A31335G
Specifications
| Amodiaquin Dihydrochloride Dihydrate | |
| Yellow | |
| MFCD00078857 | |
| amodiaquin dihydrochloride dihydrate, amodiaquine hcl, amodiaquine hydrochloride, amodiaquine dihydrochloride dihydrate, unii-k6pw2s574l, 4-7-chloroquinolin-4-yl amino-2-diethylamino methyl phenol dihydrochloride dihydrate, amodiaquine hydrochloride usp, cam-aq 1 | |
| CCN(CC)CC1=C(C=CC(=C1)NC2=C3C=CC(=CC3=NC=C2)Cl)O.O.O.Cl.Cl | |
| 464.812 | |
| CHEBI:50652 | |
| ≥98.0% (HPLC,T) |
| 6398-98-7 | |
| C20H28Cl3N3O3 | |
| 5 g | |
| YVNAYSHNIILOJS-UHFFFAOYSA-N | |
| 4-[(7-chloroquinolin-4-yl)amino]-2-(diethylaminomethyl)phenol;dihydrate;dihydrochloride | |
| 64646 | |
| 428.78 | |
| Crystalline Powder |
Chemical Identifiers
| 6398-98-7 | |
| 464.812 | |
| YVNAYSHNIILOJS-UHFFFAOYSA-N | |
| 64646 | |
| 4-[(7-chloroquinolin-4-yl)amino]-2-(diethylaminomethyl)phenol;dihydrate;dihydrochloride |
| C20H28Cl3N3O3 | |
| MFCD00078857 | |
| amodiaquin dihydrochloride dihydrate, amodiaquine hcl, amodiaquine hydrochloride, amodiaquine dihydrochloride dihydrate, unii-k6pw2s574l, 4-7-chloroquinolin-4-yl amino-2-diethylamino methyl phenol dihydrochloride dihydrate, amodiaquine hydrochloride usp, cam-aq 1 | |
| CHEBI:50652 | |
| CCN(CC)CC1=C(C=CC(=C1)NC2=C3C=CC(=CC3=NC=C2)Cl)O.O.O.Cl.Cl |
Safety and Handling
TSCA : No