missing translation for 'onlineSavingsMsg'
Learn More

Amodiaquin Dihydrochloride Dihydrate 98.0+%, TCI America™

Catalog No. p-7118232
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31335G 5 g
A313325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31335G Supplier TCI America Supplier No. A31335G
Only null left

Chemical Identifiers

CAS 6398-98-7
Molecular Formula C20H28Cl3N3O3
Molecular Weight (g/mol) 464.812
MDL Number MFCD00078857
InChI Key YVNAYSHNIILOJS-UHFFFAOYSA-N
Synonym amodiaquin dihydrochloride dihydrate, amodiaquine hcl, amodiaquine hydrochloride, amodiaquine dihydrochloride dihydrate, unii-k6pw2s574l, 4-7-chloroquinolin-4-yl amino-2-diethylamino methyl phenol dihydrochloride dihydrate, amodiaquine hydrochloride usp, cam-aq 1
PubChem CID 64646
ChEBI CHEBI:50652
IUPAC Name 4-[(7-chloroquinolin-4-yl)amino]-2-(diethylaminomethyl)phenol;dihydrate;dihydrochloride
SMILES CCN(CC)CC1=C(C=CC(=C1)NC2=C3C=CC(=CC3=NC=C2)Cl)O.O.O.Cl.Cl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 428.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Amodiaquin Dihydrochloride Dihydrate