missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiosulfate, Granular, Purified, Spectrum™ Chemical

$237.24 - $4546.79

Chemical Identifiers

CAS 7783-18-8
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18608806
View Documents
Spectrum Chemical Mfg Cor
A1273125GM
125 g Poly Bottle
Each for $237.24
Only null left
 
18608807
View Documents
Spectrum Chemical Mfg Cor
A1273500GM
500 g Poly Bottle
Each for $568.56
Only null left
 
18608808
View Documents
Spectrum Chemical Mfg Cor
A127325KG
2.5 kg Poly Bottle
Each for $2,298.15
Only null left
 
18608809
View Documents
Spectrum Chemical Mfg Cor
A127350KGBL
50 kg Fiber Drum
Each for $4,546.79
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7783-18-8

Specifications

7783-18-8
125 g
Poly Bottle
1
Purified
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.