missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thiosulfate, Granular, Purified, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Granular/Lump |
| Quantity | 125 g |
| Molecular Weight (g/mol) | 148.21 |
| Grade | Purified |
| Packaging | Poly Bottle |
| Chemical Name or Material | Ammonium Thiosulfate, Granular |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.