missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thioglycolate (ca. 60% in Water), TCI America™
Specifications
Specifications
| Color | Colorless |
| UN Number | 2922 |
| Formula Weight | 109.14 |
| Quantity | 500 g |
| Physical Form | Clear Liquid at 20°C |
| Chemical Name or Material | Ammonium Thioglycolate (ca. 60% in Water) |
Safety and Handling
| RTECSNumber | AI7525000 |
| TSCA | Yes |