missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thioglycolate (ca. 60% in Water), TCI America™

Catalog No. A221725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A221725G 25 g
A2217500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A221725G Supplier TCI America Supplier No. A221725G
Only null left

Chemical Identifiers

CAS 5421-46-5
Molecular Formula C2H7NO2S
Molecular Weight (g/mol) 109.143
MDL Number MFCD00137451
InChI Key ZZTCCAPMZLDHFM-UHFFFAOYSA-N
Synonym ammonium thioglycolate, ammonium mercaptoacetate, acetic acid, mercapto-, monoammonium salt, thiofaco a-50, ammonium thioglycollate, thioglycolic acid ammonium salt, usaf mo-2, unii-6p9e788vzz, thioglycollic acid, ammonium salt, ammonium 2-mercaptoacetate
PubChem CID 21534
IUPAC Name azanium;2-sulfanylacetate
SMILES C(C(=O)[O-])S.[NH4+]

Specifications

Color Colorless
UN Number 2922
Formula Weight 109.14
Quantity 25 g
Physical Form Clear Liquid at 20°C
Chemical Name or Material Ammonium Thioglycolate (ca. 60% in Water)
RTECSNumber AI7525000
TSCA Yes