missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thiocyanate TS, (U.S.P. Test Solution), Spectrum™ Chemical
A-311, 1762-95-4
$865.43 - $1779.14
Chemical Identifiers
| CAS | 1762-95-4 |
|---|---|
| Molecular Formula | CH4N2S |
| Molecular Weight (g/mol) | 76.12 |
| InChI Key | SOIFLUNRINLCBN-UHFFFAOYSA-N |
| IUPAC Name | ammonium cyanosulfanide |
| SMILES | [NH4+].[S-]C#N |
Description
Spectrum™ Chemical Ammonium Thiocyanate TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Chemical Identifiers
| 1762-95-4 | |
| 76.12 | |
| ammonium cyanosulfanide |
| CH4N2S | |
| SOIFLUNRINLCBN-UHFFFAOYSA-N | |
| [NH4+].[S-]C#N |
Specifications
| 1762-95-4,7732-18-5 | |
| CH4N2S | |
| [NH4+].[S-]C#N | |
| 76.12 | |
| Amber Glass Bottle | |
| Ammonium Thiocyanate TS |
| 0.1%,99.9% | |
| SOIFLUNRINLCBN-UHFFFAOYSA-N | |
| ammonium cyanosulfanide | |
| Chemical Solution | |
| 100 mL |