missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thiocyanate TS, (U.S.P. Test Solution), Spectrum™ Chemical
A-311, 1762-95-4
Supplier: Spectrum Chemical Mfg Cor A311100ML
Description
Spectrum™ Chemical Ammonium Thiocyanate TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Specifications
| Ammonium Thiocyanate TS | |
| 0.1%,99.9% | |
| SOIFLUNRINLCBN-UHFFFAOYSA-N | |
| ammonium cyanosulfanide | |
| Chemical Solution | |
| 100 mL |
| 1762-95-4,7732-18-5 | |
| CH4N2S | |
| [NH4+].[S-]C#N | |
| 76.12 | |
| Amber Glass Bottle |
Chemical Identifiers
| 1762-95-4 | |
| 76.12 | |
| ammonium cyanosulfanide |
| CH4N2S | |
| SOIFLUNRINLCBN-UHFFFAOYSA-N | |
| [NH4+].[S-]C#N |