missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate TS, 8% (w/v), Ricca Chemical

$174.10 - $174.10

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6969-16
View Documents
Ricca Chemical Company
696916
500 mL Natural Poly Bottle
Each for $174.10
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1762-95-4
76.117
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
azanium;thiocyanate
CH4N2S
SOIFLUNRINLCBN-UHFFFAOYSA-N
15666
C(#N)[S-].[NH4+]

Specifications

Colorless
91.0,7.26
CH4N2S
SOIFLUNRINLCBN-UHFFFAOYSA-N
azanium;thiocyanate
15666
Laboratory
Aqueous Solution
500 mL
8% (w/v)
1762-95-4 , 7732-18-5
94.0,7.56
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
C(#N)[S-].[NH4+]
76.117
8%
Natural Poly Bottle
4 to 7
Liquid
Ammonium Thiocyanate, Test Solution
Videos
SDS
Documents

Documents