missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, 30% (w/v), Ricca Chemical

Catalog No. p-3251314
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
69816 500 mL Natural Poly Bottle
69832 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69816 Supplier Ricca Chemical Company Supplier No. RI69816
Only null left

Chemical Identifiers

CAS 1762-95-4 , 7732-18-5
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Color Colorless
CAS Min % 75.0, 29.0
CAS Max % 78.0, 31.0
Percent Purity 30%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 4 to 7
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 30% (w/v)
Chemical Name or Material Ammonium Thiocyanate
Show More Show Less