missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiocyanate, 0.1N Standardized Solution

CAS: 7732-18-5 | NH4SCN

$108.92 - $238.32

Chemical Identifiers

CAS 7732-18-5
Molecular Formula NH4SCN
MDL Number MFCD00011428
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35580K2
View Documents
Thermo Scientific Chemicals
035580K2
1 L
Each for $108.92
Only null left
 
AA35580K7
View Documents
Thermo Scientific Chemicals
035580K7
4 L
Each for $238.32
Only null left
 
Description

Description

Ammonium thiocyanate, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7732-18-5
NH4SCN
MFCD00011428

Specifications

7732-18-5
Liquid
MFCD00011428
Miscible with water, ethanol, methanol, ammonia and acetone
76.12
1g/cm3 at 20°C
NH4SCN
Colorless
1 L
Ammonium thiocyanate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-175-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only