missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, 0.100 N (N/10), Ricca Chemical

Catalog No. R0695000500
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R0695000500 500 mL Natural Poly Bottle
69532 1 L Natural Poly Bottle
6951 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R0695000500 Supplier Ricca Chemical Company Supplier No. R0695000500A

Chemical Identifiers

CAS 1762-95-4 , 7732-18-5
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Color Colorless
CAS Min % 97.0, 0.74
CAS Max % 100.0, 0.77
Grade Laboratory
Packaging Natural Poly Bottle
pH 4 to 7
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.100Normal
Chemical Name or Material Ammonium Thiocyanate