missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfide Solution, Reagent, ACS, 20-24%, Spectrum™ Chemical

Catalog No. p-7156950
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
2.5 L
20 L
500 mL
6 x 500 mL
Packaging:
Amber Glass Bottle
Poly-Coated Glass Bottle
Steel Can
Catalog No. Quantity Packaging
18600669 100 mL Amber Glass Bottle
18600670 500 mL Amber Glass Bottle
18600672 2.5 L Poly-Coated Glass Bottle
18600671 6 x 500 mL Amber Glass Bottle
18600673 20 L Steel Can
5 options

Catalog No. 18600669

Supplier: Spectrum Chemical Mfg Cor A1404100ML

Only null left

A1404, 12135-76-1, (NH4)2S

Spectrum™ Chemical Ammonium Sulfide Solution, Reagent, ACS is an unstable salt with the formula (NH4)2S. Common uses of AMMONIUM SULFIDE are industrial applications including photo developing and textile manufacturing. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 12135-76-1 , 7732-18-5
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

CAS Max % 80%, 20%
Quantity 100 mL
UN Number UN2683
Percent Purity 20 to 24%
Grade ACS
Packaging Amber Glass Bottle
Residue after Ignition 0.04%