missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfide, Solution, 20%, Spectrum™ Chemical

$651.25 - $2379.88

Chemical Identifiers

CAS 12135-76-1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18608849
View Documents
Spectrum Chemical Mfg Cor
A1943500MLGL
500 mL Amber Glass Bottle
Each for $651.25
Only null left
 
18608848
View Documents
Spectrum Chemical Mfg Cor
A194325LTPV
2.5 L Poly-Coated Glass Bottle
Each for $2,379.88
Only null left
 
Description

Description

Specifications

Chemical Identifiers

12135-76-1

Specifications

12135-76-1 , 7732-18-5
20%
Amber Glass Bottle
500 mL
Reagent
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.