missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfide, Light Solution, Reagent, 20-24%, Spectrum™ Chemical

Catalog No. p-7156924
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
2.5 L
20 L
200 L
500 mL
6 x 500 mL
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
Catalog No. Quantity Packaging
18600674 100 mL Amber Glass Bottle
18600675 500 mL Amber Glass Bottle
18600677 2.5 L Poly-Coated Glass Bottle
18600676 6 x 500 mL Amber Glass Bottle
18600678 20 L Poly Drum
18600679 200 L Drum
6 options

Catalog No. 18600674

Supplier: Spectrum Chemical Mfg Cor A1250100ML

Only null left

A1250, 12135-76-1, (NH4)2S

Spectrum™ Chemical Ammonium Sulfide, Light Solution, Reagent , is an unstable salt with the formula (NH4)2S. It has a very strong odor, and the Reagent grade means it is used in the laboratory as an analytical testing agent. Common uses of AMMONIUM SULFIDE, LIGHT SOLUTION, REAGENT are industrial applications including photo developing, textile manufacturing and others.

Chemical Identifiers

CAS 12135-76-1 , 7732-18-5
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

CAS Max % 80%
Quantity 100 mL
UN Number UN2683
Percent Purity 20 to 24%
Grade Reagent
Packaging Amber Glass Bottle
Residue after Ignition 0.04%