missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfate, 99%, for biochemistry, Thermo Scientific™
Chemical Identifiers
| CAS | 7783-20-2 |
|---|---|
| Molecular Formula | H8N2O4S |
| Molecular Weight (g/mol) | 132.14 |
| InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
| Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
| PubChem CID | 6097028 |
| ChEBI | CHEBI:62946 |
| IUPAC Name | diazanium;sulfate |
| SMILES | [NH4+].[NH4+].[O-]S(=O)(=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC447135000
|
N/A | 500g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC447130025
|
N/A | 2.5kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 7783-20-2 | |
| 132.14 | |
| ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
| CHEBI:62946 | |
| [NH4+].[NH4+].[O-]S(=O)(=O)[O-] |
| H8N2O4S | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| 6097028 | |
| diazanium;sulfate |
Specifications
| 7783-20-2 | |
| (NH4)2SO4 | |
| ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| diazanium;sulfate | |
| 6097028 | |
| 132.14 | |
| Biochemistry |
| H8N2O4S | |
| 15,552 | |
| Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone | |
| [NH4+].[NH4+].[O-]S(=O)(=O)[O-] | |
| 132.14 | |
| CHEBI:62946 | |
| 99% | |
| Ammonium sulfate |
Safety and Handling
EINECSNumber : 231-984-1
RUO – Research Use Only