missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, 99%, for biochemistry, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 447130025

Quantity 2.5kg

 View more versions of this product

Catalog No. AC447130025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7783-20-2
132.14
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
CHEBI:62946
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
H8N2O4S
BFNBIHQBYMNNAN-UHFFFAOYSA-N
6097028
diazanium;sulfate

Specifications

Ammonium sulfate
2.5kg
(NH4)2SO4
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
BFNBIHQBYMNNAN-UHFFFAOYSA-N
diazanium;sulfate
6097028
132.14
Biochemistry
7783-20-2
H8N2O4S
15,552
Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
132.14
CHEBI:62946
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-984-1

SDS
Documents

Documents

Promotions

Promotions