missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, 2.0 M, Ricca Chemical

$157.72 - $157.72

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
694-16
View Documents
Ricca Chemical Company
69416
500 mL Natural Poly Bottle
Each for $157.72
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7783-20-2
132.13
BFNBIHQBYMNNAN-UHFFFAOYSA-N
6097028
sulfuric acid diamine
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
CHEBI:62946
N.N.OS(O)(=O)=O

Specifications

Colorless
78.0,20.5
H8N2O4S
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
N.N.OS(O)(=O)=O
132.13
CHEBI:62946
Slight ammonia oder
5.5
Liquid
7783-20-2,7732-18-5
81.0,21.33
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
sulfuric acid diamine
6097028
Laboratory
Natural Poly Bottle
500 mL
Ammonium Acetate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'