missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, 2.0 M, Ricca Chemical
Supplier: Ricca Chemical Company 69416
Specifications
| Ammonium Acetate | |
| 7783-20-2,7732-18-5 | |
| 81.0,21.33 | |
| MFCD00003391 | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| sulfuric acid diamine | |
| 6097028 | |
| Laboratory | |
| Natural Poly Bottle | |
| 500 mL |
| Colorless | |
| 78.0,20.5 | |
| H8N2O4S | |
| ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
| N.N.OS(O)(=O)=O | |
| 132.13 | |
| CHEBI:62946 | |
| Slight ammonia oder | |
| 5.5 | |
| Liquid |
Chemical Identifiers
| 7783-20-2 | |
| 132.13 | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| 6097028 | |
| sulfuric acid diamine |
| H8N2O4S | |
| MFCD00003391 | |
| ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
| CHEBI:62946 | |
| N.N.OS(O)(=O)=O |