missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, 10% (w/v), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 89.0, 9.0 |
| CAS Max % | 93.0, 11.0 |
| Percent Purity | 10% |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | 5.5 |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Concentration or Composition Notes | 10% (w/v) |
| Show More |