missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Standard, 1000 ppm NH4+ (776.5 ppm N), Ricca Chemical

Catalog No. 69316
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
69316 500 mL Natural Poly Bottle
69332 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69316 Supplier Ricca Chemical Company Supplier No. 69316

NH4Cl in Water

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.29
CAS Max % 100.0, 0.3
Equivalence 1 mL = 1 mg NH
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Ammonium Standard