missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Solution, 400 g (NH4)2HPO4 + 800 mL Water, Ricca Chemical

APHA, EPA for Sulfide Testing

$141.72 - $141.72

Chemical Identifiers

Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
690-16
SDS View Documents
Ricca Chemical Company
69016
500 mL Natural Poly Bottle
Each for $141.72
Only null left
 
Description

Description

Specifications

Chemical Identifiers

H9N2O4P
MFCD00010891
phosphoric acid diamine
132.06
MNNHAPBLZZVQHP-UHFFFAOYSA-N
N.N.OP(O)(O)=O

Specifications

7732-18-5
MFCD00010891
N.N.OP(O)(O)=O
132.06
Natural Poly Bottle
500 mL
H9N2O4P
MNNHAPBLZZVQHP-UHFFFAOYSA-N
phosphoric acid diamine
Laboratory
∼8
AMMONIUM PHOSPHATE, 400 + 800
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'