missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate, For Sulfide, 40%, LabChem™

Catalog No. LC112957
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC112957 125 mL
LC112951 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC112957 Supplier LabChem Supplier No. LC112957

Chemical Identifiers

CAS 7783-28-0 , 7732-18-5
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

Color Colorless
CAS Max % 60, 40
Linear Formula (NH4)2HPO4
Solubility Information Soluble in water
Formula Weight 132.06
Percent Purity 40%
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Gaseous Ammonia; Phosphorus oxides
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Ammonium Phosphate, For Sulfide
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature