missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Peroxydisulfate, Crystal, ACS, 98%, Spectrum™ Chemical
$207.44 - $6278.91
Chemical Identifiers
| CAS | 7727-54-0 |
|---|---|
| Molecular Formula | H8N2O8S2 |
| Molecular Weight (g/mol) | 228.19 |
| InChI Key | ROOXNKNUYICQNP-UHFFFAOYSA-N |
| IUPAC Name | diammonium [(sulfonatoperoxy)sulfonyl]oxidanide |
| SMILES | [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18608743
|
Spectrum Chemical Mfg Cor
A122525GM |
25 g | Poly Bottle |
Each for $207.44
|
|
||||
|
18608744
|
Spectrum Chemical Mfg Cor
A1225100GM |
100 g | Poly Bottle |
Each for $303.53
|
|
||||
|
18608745
|
Spectrum Chemical Mfg Cor
A1225500GM |
500 g | Poly Bottle |
Each for $412.50
|
|
||||
|
18608746
|
Spectrum Chemical Mfg Cor
A122525KG |
2.5 kg | Poly Pail |
Each for $1,345.50
|
|
||||
|
18608747
|
Spectrum Chemical Mfg Cor
A122525KGBL |
25 kg | Poly Pail |
Each for $4,181.00
|
|
||||
|
18608748
|
Spectrum Chemical Mfg Cor
A122550KGBL |
50 kg | Fiber Drum |
Each for $6,278.91
|
|
||||
Chemical Identifiers
| 7727-54-0 | |
| 228.19 | |
| diammonium [(sulfonatoperoxy)sulfonyl]oxidanide |
| H8N2O8S2 | |
| ROOXNKNUYICQNP-UHFFFAOYSA-N | |
| [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
Specifications
| 7727-54-0 | |
| 25 g | |
| ROOXNKNUYICQNP-UHFFFAOYSA-N | |
| diammonium [(sulfonatoperoxy)sulfonyl]oxidanide | |
| 98% | |
| Poly Bottle |
| 1 | |
| H8N2O8S2 | |
| [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
| 228.19 | |
| ACS | |
| 0.0005 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.