missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate TS, 3.5% (w/v), Ricca Chemical

Catalog No. p-3251301
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
67916 500 mL Natural Poly Bottle
67932 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 67916 Supplier Ricca Chemical Company Supplier No. 67916
Only null left

Meets USP TS and ACS RS specifications

Chemical Identifiers

CAS 6009-70-7 , 7732-18-5
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

CAS Min % 95.0, 3.32
CAS Max % 99.0, 3.45
Percent Purity 3.5%
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼6 to 7
Quantity 500 mL
Chemical Name or Material AMMONIUM OXALATE