missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate TS, Ricca Chemical

$234.88 - $370.81

Chemical Identifiers

CAS 7732-18-5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
674-4
SDS View Documents
Ricca Chemical Company
6744
120 mL
Each for $234.88
Only null left
 
674-16
View Documents
Ricca Chemical Company
67416
500 mL
Each for $370.81
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5

Specifications

7732-18-5,7697-37-2,1336-21-6,7782-91-4
45.0,40.32,10.94,5.45
Amber Poly Bottle
AMMONIUM MOLYBDATE
44.0,38.74,10.51,5.24
USP
120 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'