missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate Reagent II, Ricca Chemical

Catalog No. p-3251295
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
67316 500 mL Natural Poly Bottle
67332 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 67316 Supplier Ricca Chemical Company Supplier No. 67316
Only null left

For Phosphorus by the Stannous Chloride Method (without extraction)

Chemical Identifiers

CAS 12054-85-2 , 7732-18-5 , 7664-93-9
Molecular Formula H6MoNO2
Molecular Weight (g/mol) 148.00
InChI Key XCOMAVCHADVANQ-UHFFFAOYSA-O
Synonym Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate
PubChem CID 3084102
IUPAC Name ammonium hydrate molybdenum oxidandiide
SMILES [NH4+].O.[O--].[Mo]

Specifications

Color Colorless to Green-Yellow
CAS Min % 3.91, 78.0, 15.94
CAS Max % 4.07, 81.0, 16.59
Grade Laboratory
Packaging Natural Poly Bottle
pH <3
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Ammonium Molybdate Reagent II