missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Réactif Molybdate d’ammonium II, pour le phosphore, LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Ammonium Molybdate Reagent II |
| Nom Remarque | Pour le phosphore |
| Couleur | Incolore |
| CAS % maximal | 5.08, 77.86, 17.06 |
| Formule linéaire | (NH4)6Mo7O24·4H2O |
| Numéro UN | UN2796 |
| Densité | 1.1g/mL |
| Renseignements sur la solubilité | Soluble dans l’eau |
| Poids de formule | 1235.86 |
| Désignation | Test de réussite |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Wear protective gloves/eye protection. Wash exposed skin thoroughly after handling. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Température ambiante |