missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate, 4% (w/v), Ricca Chemical

Catalog No. 66532
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
66532 1 L Natural Poly Bottle
66516 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 66532 Supplier Ricca Chemical Company Supplier No. 66532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12054-85-2 , 7732-18-5

Specifications

Color Colorless
CAS Min % 94.0, 3.0
CAS Max % 98.0, 5.0
Percent Purity 4%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 5 to 5.5
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 4% (w/v)
Chemical Name or Material Ammonium Molybdate
Show More Show Less