missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) sulfate dodecahydrate, MilliporeSigma™

For analysis EMSURE™ ACS, ISO, Reag. Ph Eur

Supplier:  Technidata 1037765000

 View more versions of this product

Catalog No. M1037765000


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ammonium iron(III) sulfate dodecahydrate
5 kg
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
LCPUDZUWZDSKMX-UHFFFAOYSA-K
azanium;iron(3+);disulfate;dodecahydrate
197097
Odorless
7783-83-7
FeH28NO20S2
Soluble in water: 1240g/L at 25°C
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
482.176
482.19
Plastic Bottle

Chemical Identifiers

7783-83-7
482.176
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
azanium;iron(3+);disulfate;dodecahydrate
FeH28NO20S2
LCPUDZUWZDSKMX-UHFFFAOYSA-K
197097
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-382-4

Recommended Storage : Store below 15°C (59°F), Tightly closed. Dry. Protected from Light.

SDS
Documents

Documents

Promotions

Promotions