missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium iron(III) sulfate dodecahydrate, MilliporeSigma™
For analysis EMSURE™ ACS, ISO, Reag. Ph Eur
$307.01 - $9243.07
Chemical Identifiers
| CAS | 7783-83-7 |
|---|---|
| Molecular Formula | FeH28NO20S2 |
| Molecular Weight (g/mol) | 482.176 |
| InChI Key | LCPUDZUWZDSKMX-UHFFFAOYSA-K |
| Synonym | ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g |
| PubChem CID | 197097 |
| IUPAC Name | azanium;iron(3+);disulfate;dodecahydrate |
| SMILES | [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1037761000
|
Technidata
1037761000 |
1 kg | Plastic Bottle |
Each for $307.01
|
|
||||
|
M1037765000
|
Technidata
1037765000 |
5 kg | Plastic Bottle |
Each for $856.11
|
|
||||
|
M1037769050
|
Technidata
1037769050 |
50 kg | Fibre carton |
Each for $9,243.07
|
|
||||
Chemical Identifiers
| 7783-83-7 | |
| 482.176 | |
| ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
| azanium;iron(3+);disulfate;dodecahydrate |
| FeH28NO20S2 | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| 197097 | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
Specifications
| 7783-83-7 | |
| FeH28NO20S2 | |
| Soluble in water: 1240g/L at 25°C | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] | |
| 482.176 | |
| 482.19 | |
| Plastic Bottle |
| 1 kg | |
| ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| azanium;iron(3+);disulfate;dodecahydrate | |
| 197097 | |
| Odorless | |
| Ammonium iron(III) sulfate dodecahydrate |
Safety and Handling
EINECSNumber : 233-382-4
Recommended Storage : Store below 15°C (59°F), Tightly closed. Dry. Protected from Light.