missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 25% v/v, Reagents
Ammonium Hydroxide, 25% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CA1003004A
Specifications
| >11.5 | |
| Colorless | |
| 7732-18-5,1336-21-6 | |
| Aqua Ammonia | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 25% | |
| Poly Bottle | |
| Liquid |
| Ammonium Hydroxide, 25% v/v | |
| 36°C | |
| H2O | |
| 4 L | |
| O | |
| 18.02 | |
| Reagent | |
| 0.975 |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Aqua Ammonia | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |