missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 25% v/v, Reagents
Specifications
Specifications
| pH | >11.5 |
| Color | Colorless |
| Boiling Point | 36°C |
| Quantity | 1 L |
| Percent Purity | 25% |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 0.975 |
| Physical Form | Liquid |
| Chemical Name or Material | Ammonium Hydroxide, 25% v/v |