missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 20% (v/v), Ricca Chemical

Supplier:  Ricca Chemical Company RI6351

 View more versions of this product

Catalog No. 635-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Hydroxide
1336-21-6,7732-18-5
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
19.3 to 20.7%
Natural Poly Bottle
Liquid
Colorless
H5NO
20% (v/v)
4 L
N.O
35.05
CHEBI:18219
Laboratory
Aqueous Solution

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
amine hydrate
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
N.O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions