missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, Puriss. p.a., ACS Reagent, ≥98%, Honeywell Fluka™

Puriss. p.a., ACS Reagent,≥98%

Supplier:  Honeywell-Fluka 301011KG

 View more versions of this product

Catalog No. 6000334


Only null left
Explore available promotions

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Ammonium Fluoride
1 kg
NH4F
2505
N.F
37.04
≥98%
Plastic Bottle
12125-01-8
FH4N
MFCD00011423
LDDQLRUQCUTJBB-UHFFFAOYSA-N
amine hydrofluoride
37.04g/mol
ACS/puriss. p.a.
≤0.005% (as SO4)

Chemical Identifiers

12125-01-8
37.04
LDDQLRUQCUTJBB-UHFFFAOYSA-N
N.F
FH4N
MFCD00011423
amine hydrofluoride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P280-P302 + P352 + P312-P304 + P340 + P312-P305 + P351 + P338 + P310

H301 + H311 + H331-H318

EINECSNumber : 235-185-9

SDS
Documents

Documents

Promotions

Promotions