missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, Puriss. p.a., ACS Reagent, ≥98%, Honeywell Fluka™

Catalog No. p-7071958
Change view
Click to view available options
Quantity:
100 g
1 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000333 100 g
6000334 1 kg
60045801 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000333 Supplier Honeywell Chemicals Supplier No. 30101100G
Only null left

Puriss. p.a., ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Quantity 100 g
Linear Formula NH4F
UN Number 2505
Formula Weight 37.04g/mol
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Packaging Plastic Bottle
Residue after Ignition ≤0.005% (as SO4)
Chemical Name or Material Ammonium Fluoride
Hazard Statement P280-P302 + P352 + P312-P304 + P340 + P312-P305 + P351 + P338 + P310
Precautionary Statement H301 + H311 + H331-H318
EINECSNumber 235-185-9