missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Fluoride, Crystal, ACS, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor A11851KG

 View more versions of this product

Catalog No. 18608646


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

12125-01-8
1 kg
ACS
0.0001
1
98%
Poly Bottle

Chemical Identifiers

12125-01-8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.