missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Fluoride, Crystal, ACS, 98%, Spectrum™ Chemical

Catalog No. p-7164989
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
1 kg
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Catalog No. Quantity Packaging
18608647 25 g Poly Bottle
18608641 125 g Poly Bottle
18608643 500 g Poly Bottle
18608646 1 kg Poly Bottle
18608645 2.5 kg Poly Bottle
18608642 12 kg Fiber Drum
18608644 50 kg Fiber Drum
7 options

Catalog No. 18608647

Supplier: Spectrum Chemical Mfg Cor A118525GM

Only null left

Chemical Identifiers

CAS 12125-01-8

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98%
Grade ACS
Packaging Poly Bottle
Residue after Ignition 0.0001

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.