missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate Solution R, 158 g/L, Ricca Chemical

Catalog No. 6221516
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6221516 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6221516 Supplier Ricca Chemical Company Supplier No. 622.1516

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 506-87-6 , 7732-18-5
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Color Colorless
CAS Min % 85.0, 13.38
CAS Max % 88.0, 13.92
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼9
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 158g/L
Chemical Name or Material Ammonium Carbonate Solution, Reagent