missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia TS 2, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Odor | Characteristic ammonia odor |
| Assay Percent Range | 13.5 ± 0.5% (v/v) |
| Physical Form | Liquid |
| Color | Colorless |
| pH | Alkaline |
| Melting Point | Approximately 0°C |
| Boiling Point | Approximately 36°C |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Solubility | Infinitely soluble in water |
| Show More |