missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Standard, Certified, 1000ppm ±10ppm (1mL = 1mg NH3), LabChem™

Catalog No. LC109001
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC109001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC109001 Supplier LabChem Supplier No. LC109001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Packaging Poly Bottle
CAS Max % 99.69, 0.31
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg NH3)
Linear Formula NH4Cl
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 53.49
Grade Certified
Assay as NH3
Chemical Name or Material Ammonia Standard, 1000ppm (1mL = 1mg NH3)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature