missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Standard, 10 ppm NH3 (8.22 ppm N), Ricca Chemical

Catalog No. p-3251256
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6124 120 mL Natural Poly Bottle
61216 500 mL Natural Poly Bottle
61232 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6124 Supplier Ricca Chemical Company Supplier No. 6124
Only null left

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.0030
CAS Max % 100.0, 0.0032
Equivalence 1 mL = 0.01 mg NH3
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Ammonia Standard