missing translation for 'onlineSavingsMsg'
Learn More

Ammonia (as Nitrogen) Standard, Certified, 1000ppm ±10ppm (1mL = 1mg N, 1mL = 1.22mg NH3), LabChem™

Catalog No. p-5379542
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC179407 125 mL Poly Bottle
LC179401 500 mL Poly Bottle
LC179402 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC179407 Supplier LabChem Supplier No. LC179407
Only null left

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Packaging Poly Bottle
CAS Max % 99.62, 0.38
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg N, 1mL = 1.22mg NH3)
Linear Formula NH4Cl
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 53.49
Grade Certified
Assay as N
Chemical Name or Material Ammonia (as Nitrogen) Standard, 1000ppm, (1mL = 1mg N, 1mL = 1.22mg NH3)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature