missing translation for 'onlineSavingsMsg'
Learn More

Ammonia (as Nitrogen) Standard, Certified, 1000ppm ±10ppm (1mL = 1mg N, 1mL = 1.22mg NH3), LabChem™

Supplier:  LabChem LC179402

 View more versions of this product

Catalog No. LC179402


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ammonia (as Nitrogen) Standard
Poly Bottle
99.62,0.38
1g/mL
as N
1000 ppm ±10 ppm (1mL = 1mg N, 1mL = 1.22mg NH3)
MFCD00011420
Soluble in water
N.Cl
53.49
25517
53.49
1000ppm, (1mL = 1mg N, 1mL = 1.22mg NH3)
12125-02-9,7732-18-5
Passes Test
Liquid
ClH4N
NH4Cl
Colorless
NLXLAEXVIDQMFP-UHFFFAOYSA-N
amine hydrochloride
1 L
CHEBI:31206
Certified

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
CHEBI:31206
N.Cl
ClH4N
MFCD00011420
25517
amine hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions