missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia, 0.5M solution in 1,4-dioxane, AcroSeal™
CAS: 7664-41-7 | H3N | 17.03 g/mol
$193.07 - $193.07
Specifications
| Chemical Name or Material | Ammonia |
|---|---|
| Color | Colorless |
| Name Note | 0.5M solution in 1, 4-dioxane |
| CAS | 7664-41-7 , 123-91-1 |
| Molecular Formula | H3N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC368381000
|
Thermo Scientific Chemicals
368381000 |
100 mL | AcroSeal™ Glass bottle |
Each for $193.07
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ammonia | |
| 0.5M solution in 1, 4-dioxane | |
| H3N | |
| MFCD00011418 | |
| 05,15 | |
| 15, 488 | |
| Solubility in water: soluble. | |
| N | |
| 222 | |
| 17.03 | |
| 11°C |
RUO – Research Use Only