missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amisulpride 98.0+%, TCI America™
$275.87 - $1204.17
Chemical Identifiers
| CAS | 71675-85-9 |
|---|---|
| Molecular Formula | C17H27N3O4S |
| Molecular Weight (g/mol) | 369.48 |
| MDL Number | MFCD00866691 |
| InChI Key | NTJOBXMMWNYJFB-UHFFFAOYSA-N |
| Synonym | amisulpride, aminosultopride, solian, amisulprida, amisulpridum, deniban, amisulpridum inn-latin, amisulprida inn-spanish, amisulpride inn, amisulpiride |
| PubChem CID | 2159 |
| ChEBI | CHEBI:64045 |
| IUPAC Name | 4-amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-ethylsulfonyl-2-methoxybenzamide |
| SMILES | CCN1CCCC1CNC(=O)C2=CC(=C(C=C2OC)N)S(=O)(=O)CC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A2450100MG
|
TCI America
A2450100MG |
100 mg |
Each for $275.87
|
|
|||||
|
A24501G
|
TCI America
A24501G |
1 g |
Each for $1,204.17
|
|
|||||
Chemical Identifiers
| 71675-85-9 | |
| 369.48 | |
| NTJOBXMMWNYJFB-UHFFFAOYSA-N | |
| 2159 | |
| 4-amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-ethylsulfonyl-2-methoxybenzamide |
| C17H27N3O4S | |
| MFCD00866691 | |
| amisulpride, aminosultopride, solian, amisulprida, amisulpridum, deniban, amisulpridum inn-latin, amisulprida inn-spanish, amisulpride inn, amisulpiride | |
| CHEBI:64045 | |
| CCN1CCCC1CNC(=O)C2=CC(=C(C=C2OC)N)S(=O)(=O)CC |
Specifications
| 71675-85-9 | |
| White | |
| MFCD00866691 | |
| amisulpride, aminosultopride, solian, amisulprida, amisulpridum, deniban, amisulpridum inn-latin, amisulprida inn-spanish, amisulpride inn, amisulpiride | |
| CCN1CCCC1CNC(=O)C2=CC(=C(C=C2OC)N)S(=O)(=O)CC | |
| 369.48 | |
| CHEBI:64045 | |
| ≥98.0% (HPLC,T) | |
| Amisulpride |
| 127°C | |
| C17H27N3O4S | |
| 100 mg | |
| NTJOBXMMWNYJFB-UHFFFAOYSA-N | |
| 4-amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-ethylsulfonyl-2-methoxybenzamide | |
| 2159 | |
| 369.48 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CV2308701
TSCA : No
Recommended Storage : Refrigerator