accessibility menu, dialog, popup

UserName

CAS RN 71675-85-9

CH3NNHOH2NOCH3SOOH3C

CAS RN 71675-85-9

IUPAC Name: 4-amino-5-(ethanesulfonyl)-N-[(1-ethylpyrrolidin-2-yl)methyl]-2-methoxybenzamide
Synonyms: amisulpride sultroprida 氨磺必利 amisulpride - chemical solian barnetil amisulprida topral amisulpride (product) sultopridum
Molecular Weight (g/mol): 369.48
Molecular Formula: C17H27N3O4S
InChi Key: NTJOBXMMWNYJFB-UHFFFAOYNA-N
SMILES: CCN1CCCC1CNC(=O)C1=CC(=C(N)C=C1OC)S(=O)(=O)CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results