missing translation for 'onlineSavingsMsg'
Learn More

Aminopterin Hydrate 97.0+%, TCI America™

Supplier:  TCI America A0410100MG

 View more versions of this product

Catalog No. A0410100MG

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Aminopterin Hydrate
222°C
C19H20N8O5
100 mg
4-Aminofolic Acid, 4-Amino-PGA, 4-Aminopteroyl-L-glutamic Acid
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=NC(=N3)N)N
440.42
CHEBI:22526
≥97.0% (HPLC,T)
54-62-6
Yellow
MFCD00036692
2811
TVZGACDUOSZQKY-LBPRGKRZSA-N
(2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
169371
440.42
Crystalline Powder

Chemical Identifiers

54-62-6
440.42
TVZGACDUOSZQKY-LBPRGKRZSA-N
169371
(2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
C19H20N8O5
MFCD00036692
4-Aminofolic Acid, 4-Amino-PGA, 4-Aminopteroyl-L-glutamic Acid
CHEBI:22526
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=NC(=N3)N)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : MA1050000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions